Jerricans, electroconductive, HDPE
Leverandør: BURKLE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
0435-0005 0435-0015 0435-1055 0435-1105 0435-2055 0435-2105
216-2227EA 1570 DKK
216-2227 216-2228 216-2229 216-2230 216-2332 216-2333
Jerricans, electroconductive, HDPE
Flasker Metaldunke
HDPE, black, anti-static.
  • With or without threaded connector (³/₄")
  • Ribbed structure and thick sides make it very stable and also allow for flat storage
  • Handle ensures secure handling and easy transport
  • Due to its narrow width (65 or 125 mm), this space-saving canister requires very little room, making it suitable for use in very confined spaces
This compact jerrican, made of electrically conductive HDPE, is ideal for use in explosion proof areas. With a specific surface resistance of <10⁵ ohms, the canister has a very high electrical conductivity.More Product Information