Electrolyte solutions, aqueous
Leverandør: SI Analytics
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
285136923 285138349 285138649 285138151 285138560 285138590
SCOI285136923EA 468 DKK
SCOI285136923 662-6184 SCOI285138649 SCOI285138151 SCOI285138560 SCOI285138590
Electrolyte solutions, aqueous
Elektrolytter Opløsninger til Electrodefyldning
Electrolyte aqueous solutions are used for reference electrodes, as electrolyte bridges and for storage.