ESD safety shoes, lace-up, uvex 1 sport, 6596
Leverandør: UVEX
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
6596.2/35 6596.2/36 6596.2/37 6596.2/45 6596.2/46 6596.2/49 6596.2/50 6596.2/41 6596.2/38 6596.2/42 6596.2/51 6596.2/48 6596.2/43 6596.2/44 6596.2/47 6596.2/39 6596.2/40 6596.2/52 6596.3/41 6596.3/35 6596.3/51 6596.3/44 6596.3/48 6596.3/47 6596.3/40 6596.3/50 6596.3/37 6596.3/39 6596.3/42 6596.3/38 6596.3/45 6596.3/46 6596.3/36 6596.3/49 6596.3/52 6596.3/43 6596.1/35 6596.1/36 6596.1/37 6596.1/38 6596.1/39 6596.1/40 6596.1/41 6596.1/42 6596.1/43 6596.1/44 6596.1/45 6596.1/46 6596.1/47 6596.1/48 6596.1/49 6596.1/50 6596.1/51 6596.1/52
114-8552EA 812 DKK
114-8552 114-8553 114-8554 114-8562 114-8563 114-8566 114-8567 114-8558 114-8555 114-8559 114-8568 114-8565 114-8560 114-8561 114-8564 114-8556 114-8557 114-8569 UVEX6596.3/41 UVEX6596.3/35 UVEX6596.3/51 UVEX6596.3/44 UVEX6596.3/48 UVEX6596.3/47 UVEX6596.3/40 UVEX6596.3/50 UVEX6596.3/37 UVEX6596.3/39 UVEX6596.3/42 UVEX6596.3/38 UVEX6596.3/45 UVEX6596.3/46 UVEX6596.3/36 UVEX6596.3/49 UVEX6596.3/52 UVEX6596.3/43 UVEX6596.1/35 UVEX6596.1/36 UVEX6596.1/37 UVEX6596.1/38 UVEX6596.1/39 UVEX6596.1/40 UVEX6596.1/41 UVEX6596.1/42 UVEX6596.1/43 UVEX6596.1/44 UVEX6596.1/45 UVEX6596.1/46 UVEX6596.1/47 UVEX6596.1/48 UVEX6596.1/49 UVEX6596.1/50 UVEX6596.1/51 UVEX6596.1/52
ESD safety shoes, lace-up, uvex 1 sport, 6596
Sko og støvler
Metal-free, extremely lightweight, flexible, multifunctional safety shoe made from breathable perforated high-tech micro velour upper, soft padded collar and dust tongue, plus a dual-density PUR sole (oil and petrol resistant, anti-static) free of paint wetting inhibitors such as silicones, plasticisers and phthalates.
  • Removable anti-static comfort insole with moisture transport system; orthopaedic insoles possible
  • Breathable distance mesh lining
  • Anti-slip sole with shock-absorbent heel and front of the foot
  • Virtually seamless design eliminates potential pressure points
  • 100% metal-free xenova® toecap, non magnetic (200 J)
  • Fulfils ESD requirements: Resistance to ground inferior to 35 megaohms
Suitable for people allergic to chrome.
Colour: Black
Width: 10, 11, 12
Certificeringer: EN 61340 (ESD); DGUV112-191; EN ISO 20345:2011; S3, SRC