Leather gloves, uvex top grade 8400
Leverandør: UVEX
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
6029108 6029109 6029110 6029111 6029112
UVEX6029108EA 915 DKK
UVEX6029108 UVEX6029109 UVEX6029110 UVEX6029111 UVEX6029112
Leather gloves, uvex top grade 8400
Handsker
These safety gloves are made of extremely high quality leather with cotton on palm.
  • Fingertip, wrist and knuckle protection
  • Good mechanical abrasion resistance
  • Exceptional grip on dry and (slightly) damp tools
  • Excellent wearer comfort
The glove is constructed of 100% full-grain leather, cuff and internal elastic collar on the back of the hand.
Typical applications include: Assembly, inspection, light to medium metal processing and manual work.
Certificeringer: EN 388:2016 (3132X)
Bestil nu
Specifikationer
Model 8400
Ambidextrous/Hånd-specifik Hånd-specifik
Environmentally Preferable
Engangs/Genbrugelig Genanvendelig
Manchet Elastisk
EN norm EN 388:2016 (3132X)