Industrial gloves, uvex phynomic x-foam HV
Leverandør: UVEX
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
6005406 6005407 6005408 6005409 6005410
UVEX6005406EA 550 DKK
UVEX6005406 UVEX6005407 UVEX6005408 UVEX6005409 UVEX6005410
Industrial gloves, uvex phynomic x-foam HV
Handsker
These polyamide/elastane safety gloves with aqua-polymer foam coating on palm and fingertips fit like a second skin and are particularly light and flexible, without making the hands sweat.
  • Reduced tear resistance in the fingers due to seamless break sections, significantly reducing the risk of hand injuries
  • Very good grip in dry and slightly damp areas
  • Outstanding tactile feel when assembling parts
  • Highly breathable coating with good mechanical abrasion resistance
  • Very good skin compatibility
Suitable for use when handling moving machine parts.