ESD safety ankle boots, lace-Up, L10, shadow mid
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
761081/37 761081/39 761081/40 761081/41 761081/42 761081/44 761081/45 761081/46 761081/47
114-8800EA 701 DKK
114-8800 114-8802 114-8803 114-8804 114-8805 114-8807 114-8808 114-8809 114-8810
ESD safety ankle boots, lace-Up, L10, shadow mid
Sko og støvler
Casual, vintage style, environmental friendly manufactured ESD safety ankle boot made from cowhide leather, a full padded bellows tongue, leather lining and a dual density TPU/PU sole (L10).
  • Very low weight helps to lightweight upper and sole materials
  • Anatomically shaped insole and ankle support for improved stability
  • Replaceable insole; orthopaedic insoles possible
  • Full-length inlay sole ESD PRO beige
  • Metal-free, puncture resistant midsole
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black
Certificeringer: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC