Industrial gloves, uvex profi ergo, ENB20A
Leverandør: UVEX
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
6014700 6014701 6014704 6014702 6014703
UVEX6014700EA 431 DKK
UVEX6014700 UVEX6014701 UVEX6014704 UVEX6014702 UVEX6014703
Industrial gloves, uvex profi ergo, ENB20A
Handsker
These cotton interlock safety gloves feature a special NBR coating on palm, fingers and ³/₄ of the back of the hand. Ideal for use in oily and greasy areas of application.
  • Very good grip in damp, wet and oily areas
  • Good tactile feel
  • Ergonomic fit, highly flexible
  • Also suitable for operating touch displays
  • Lining provides high wearer comfort, feels pleasant against the skin, and provides optimum regulation of the glove climate
Typical applications include: Assembly, inspection, repair work, shipping/logistics and packaging.
Certificeringer: EN 388:2016 (2121X)
OEKO-TEX® Standard 100