Pipette tips, gel loading, universal fit
Leverandør: GILSON
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
F1733481 F1731181 F1731281 F1733181 F1733381 F1733281 F1731182 F1731282 F1733182 F1733382 F1733282
GILSF1733481EA 396 DKK
GILSF1733481 GILSF1731181 GILSF1731281 GILSF1733181 GILSF1733381 GILSF1733281 GILSF1731182 GILSF1731282 GILSF1733182 GILSF1733382 GILSF1733282
Pipette tips, gel loading, universal fit
Pipettespidser
Tips allow you to stock one brand of tip with practical packaging options so you can save time, money, and bench space.
  • Manufactured in a sterile and clean-room environment
  • Graduations at varying volumes
  • Economical and easy to use
  • Colour coded tips
Sterilised EXPERT tips are free of human DNA, RNase, DNase, PCR inhibitors and pyrogens. This inert polymer will not leach into or degrade your samples so there’s no concern of contamination.
EXPERT filter tips feature a patented, low-retention polymer that creates a hydrophobic surface to minimize sample binding when dispensing.