Cleanroom hood system, ViVi-CR
Leverandør: THI Total Healthcare Innovation GmbH
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
80100 80201
111-2417EA 12840 DKK
111-2417 111-2418
Cleanroom hood system, ViVi-CR
Åndedrætsværn Åndedrætsbeskyttelsessystemer
Sterile full headcover system with active air filtration to protect your cleanroom and provide user comfort.
  • Two fans ensure cool and comfortable climate
  • 370 g headtop due to the use of carbon reinforced plastics
  • Performance filter to retain particles and allow air exchange
  • Clear PC visor providing 190° viewing angle
  • Gesture control for hands-free change of fan speed
  • Automatically adjusts fan speed to your level of physical activity
  • Sterility assurance level (SAL) of 10-6 (hood)
Ideal for use in sterile cleanroom production and research, where effective particle retention as much as a comfortable and non distractive user experience is required.
Suitable for use in GMP Class A/B environments.
Certificeringer: EN 166 F.
Information om tilbehør: ViVi-CR comfort pads are sold as a kit to exchange all pads on the ViVi-CR helmet.
Bestillingsoplysninger: The ViVi-CR helmet provides air exchange and positions the single use, sterile ViVi-CR hood. The ViVi-CR battery pack powers the helmet, and the ViVi-CR battery charger charges up to 4 battery packs at the same time.
Emballering: The sterile ViVi-CR hood comes in a sterile single header bag, 20 hoods are packed into a outer header bag and delivered in a carton box.