BOD measurement, OxiTop®-i with graphic display
Leverandør: WTW
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
208260 208262 208264 208266 208270 208272 208274 208276 208278
705-1144EA 32080 DKK
705-1144 705-1145 705-1148 705-1149 705-1150 705-1151 705-1152 705-1146 705-1153
BOD measurement, OxiTop®-i with graphic display
BOD bestemmelse
The OxiTop®-i is a modern, easy to use instrument for BOD measurement (Biochemical Oxygen Demand). The duration can be defined between 1 and 7 days. The sample volume can be selected on the menu. The conversion into the corresponding BOD value will be automatic, error-free and convenient. A small LED show the actual operating condition of the head. The heads have a modern design, are available in two different colours with adjustable numeric sample ID allowing distinction between inlet and outlet.
  • No need to convert data - direct input of sample volume (select from 22,7 / 43,5 / 97,0 / 164 / 250 / 365 / 432 ml)
  • Most reliable results - displays the curve at the head
  • Full information - interim values always available
  • Pressure range: 500 to 1250 hPa
  • Measurement period: 1 to 7 days
  • BOD measuring range: 0 to 4000 mg/L
The modern backlit graphic LCD supports the input and provides various information:
• Display of the curve
• Selection of sample volume
• Call-up of interim values
Sets are available which include stirring platforms and accessories:
• 6 or 12 measuring places
• Inductive stirring platforms with program controlled operation
• Two overflow measuring flasks for inlet and outlet measurement