Cut resistant gloves, Showa 4561
Leverandør: Showa
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
4561XXL-10 4561S-06 4561L-08 4561M-07 4561XL-09
112-5842EA 1670 DKK
112-5842 112-5840 112-5838 112-5839 112-5841
Cut resistant gloves, Showa 4561
Handsker
These hand-specific gloves with nitrile coating are engineered to endure as they combine Kevlar® cut-resistance with oil grip to deliver cut, abrasion and flame resistance, without sacrificing comfort or grip.
  • Sponge nitrile coating protects the hand from penetration of oils, hydrocarbons and grease, provides grip performance in oily circumstances with high abrasion resistance
  • Flame resistance level 2 (up to 250 °C approx. 15 seconds)
  • Lightweight, 15-gauge breathable liner for dry and comfortable hands
  • Seamless knit to prevent irritation
  • Excellent level of dexterity and tactility thanks to stretchy spandex fibres in the liner
  • Light, elastic low-lint, resistant to deformation
  • Designed for easy movement and continuous wear
Ideal for automotive, construction, glass and glazing, logistics, mechanical and metallurgy industries. Typical applications include: Automotive assembly, goods handling, handling components with sharp edges, handling oily components, maintenance.
Certificeringer: EN 388:2016 (4X32D); Cat. II