Pierce™ streptavidin
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
21122 21125 21135 21145
PIER21122EA 1510 DKK
PIER21122 PIER21125 PIER21135 PIER21145
Pierce™ streptavidin
Proteiner
Pierce™ Streptavidin is a purified unconjugated recombinant protein.
  • Recombinant streptavidin high-quality protein
  • Used for biotin blocking and controls
  • Mostly used in ELISA (assay dependent) and western blot (assay dependent)
Purified recombinant streptavidin enables detection of biotinylated antibodies and other probes in a variety of standard assay methods, including western blotting, ELISA, immunohistochemistry, and fluorescence imaging. Purified streptavidin is supplied lyophilised in phosphate-based buffer for immediate reconstitution with water.
Streptavidin is a biotin-binding protein originally isolated from Streptomyces avidinii. In contrast to avidin, streptavidin has no carbohydrate and has a mildly acidic pI of 5. These products use a recombinant form of streptavidin having a mass of 53000 daltons and a near-neutral pI (6,8 - 7,5). Tetrameric protein, with each subunit binding one molecule of biotin with affinity similar to that of avidin. Guanidinium chloride will dissociate avidin and streptavidin into subunits, but streptavidin is more resistant to dissociation.
Emballering: For research use only. Not for use in diagnostic procedures.