General purpose gloves, Showa 310
Leverandør: Showa
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
310OR-10/XL 310G-6/XS 310G-7/S 310G-8/M 310OR-8/M 310G-9/L 310OR-9/L 310G-10/XL 310B-10/XL 310B-7/S 310B-8/M 310B-9/L 310OR-7/S
112-5776EA 496 DKK
112-5776 112-5772 112-5773 112-5774 112-5778 112-5775 112-5779 112-5771 112-5767 112-5768 112-5769 112-5770 112-5777
General purpose gloves, Showa 310
Handsker
These hand-specific cotton gloves with latex coating on palm and fingertips are flexible and robust offering great dexterity, outstanding grip and good resistance to tearing. They are ideal for heavy duty manual handling.
  • Latex coating provides protection against abrasions, alcohols and aggressive detergents
  • Excellent grip in damp and dry environments
  • Seamless knit to prevent irritation
  • Breathable back of hand to reduce perspiration
  • Designed for easy movement and continuous wear thanks to renowned ergonomic shaped hand mould
Ideal for use in construction, highway maintenance, logistics, municipal services, heavy engineering. Typical applications include: Bricklaying, maintenance, manual handling, refuse collection and tiling.
Certificeringer: EN 388:2016 (2142X); Cat. II.