Protein A/G magnetic agarose beads, Pierce™
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
78609 78610
PIER78609EA 4190 DKK
PIER78609 PIER78610
Protein A/G magnetic agarose beads, Pierce™
Kugler Magnetiske perler
Protein A/G magnetic agarose beads provide a fast, convenient method for purification of immunoglobulins from serum, cell culture supernatant, or ascites.
  • Sufficient for both routine and demanding purification procedures
  • Optimised for use with high-throughput magnetic platforms
  • Flexible for IgG binding domains of both Protein A and Protein G on one bead
  • Beads are compatible with manual and automated applications
These beads are a mixture of Protein A magnetic beads and Protein G magnetic beads. The recombinant chimeric Protein A/G combines four IgGMore Product Information