Masking tapes, 501E
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5017250 5011850 5012450 5013650 5014850
MMMA5017250EA 2520 DKK
MMMA5017250 MMMA5011850 MMMA5012450 MMMA5013650 MMMA5014850
Masking tapes, 501E
Tape
These masking tapes have easy processing with all standard paint systems and meets the high demands of the automotive industry.
  • High temperature resistant up to 160 °C
  • No flaking off of paints
  • Solvent and moisture resistant
  • Removable without residue at one piece
  • Sharp colour edges
These tapes are suitable for sensitive painting work especially in the automotive sector, oven drying, IR and heat lamps systems.
These tapes are cuddly, paper carrier with rubber resin adhesive and has good paint and filler adhesion.
These tapes are of 50 m length and 0,15 mm thickness.