ESD safety boots, lace-up, Johnny
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
76591/38 76591/39 76591/40 76591/41 76591/43 76591/44 76591/48
ELTN76591/38EA 1640 DKK
ELTN76591/38 ELTN76591/39 ELTN76591/40 ELTN76591/41 ELTN76591/43 ELTN76591/44 ELTN76591/48
ESD safety boots, lace-up, Johnny
Sko og støvler
Extrawide ESD safety boots made from cowhide leather with full, padded bellows, collar padding tongue, reflective materials and a SAFEGUARD deep-treaded double-density PU sole with profile (4,6 mm, abrasion-, heat-, oil and fuel resistant; temperature resistant from -20 to approx. 130 °C).
  • Warm lining provides good ventilation and high moisture absorbance
  • Heel pocket lining made from sturdy abrasion resistant microfibre
  • Heel loop to pull on boots easily
  • PU scuff cap
  • Therm-ic heat control keeps the feet warm and controls the temperature inside the shoe
  • Heating element can be adjusted via an app (Bluetooth connection) and can be set to either durable heat supply or heat in intervals
  • Protective toe cap made from aluminium (200 J)
  • Metal-free puncture resistant midsole
Ideal for cold areas, working in winter or road construction.
Certificeringer: 61340-5-1 (ESD); EN ISO 20345 S3, SRC, CI
Leveringsinformation: Battery for heating element (3.7 V / 2.600 mAh) and USB charging cable are included.