VWR® VisiLight® LED70, High Power Cold Light Source without Fan
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
631-1249EA 15450 DKK
631-1249 631-1254
VWR® VisiLight® LED70, High Power Cold Light Source without Fan
Lyskilder Koldlyskilder
This fanless cold light source with passive cooling system and compact and stable aluminium housing provides an optimal and homogenous illumination of the light guide. It is therefore ideal for high speed recordings in a variety of applications, such as quality assurance, industrial endoscopy, machine vision, and cleanroom applications. The brightness exceeds by far the brightness of a conventional 250 W halogen light source. Expected LED lifespan of 50000 hours.
  • 1500 lm at the end of the light guide
  • Fanless, passive cooling system, for a noiseless and vibration-free operation
  • No contamination of the cooling system
  • Analogue brightness control from 0 to 100% (flicker-free)
  • Blinding protection by light guide detection
  • Triple-thermal fuse
  • Integrated power supply
  • Optional handle for more flexibility
W×D×H: 230×283×116 mm
Weight: 5,2 kg
* lm = luminous flux (light guide Ø = 9 mm and L = 1 m)