VWR® VisiLight® LED8, High Power Cold Light Source
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
631-1258EA 9850 DKK
631-1258 631-1262
VWR® VisiLight® LED8, High Power Cold Light Source
Lyskilder Koldlyskilder
This cold light source with compact and stable aluminium housing and temperature fan control provides an optimal and homogenous illumination of the light guide. It is therefore ideal for high speed recordings in a variety of applications, such as quality assurance, industrial endoscopy, machine vision, and cleanroom applications. The brightness exceeds by far the brightness of a conventional 250 W halogen light source. Expected LED lifespan of 50000 hours.
  • 1600 lm at the end of the light guide
  • Temperature fan control for minimal noise disruption
  • Analogue brightness control from 0 to 100% (flicker-free)
  • Blinding protection by light guide detection
  • Triple-thermal fuse
  • Integrated power supply
  • Stackable and space-saving
W×D×H: 200×273×116 mm
Weight: 2,3 kg
* Luminous flux (Light guide Ø = 9 mm and L = 1 m)