Autoclavable cleanroom goggles, 619
Leverandør: UNIVET
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
619.04.23.10 619.05.23.10
UNVT619.04.23.10EA 2580 DKK
UNVT619.04.23.10 UNVT619.05.23.10
Autoclavable cleanroom goggles, 619
Beskyttelsesbriller
These cleanroom googles are available with a direct or an indirect ventilation system. Ideal for use in rooms with contamination control (clean and white room) as well as in aseptic environments.
  • Polycarbonate lenses protect against impacts and UV radiations
  • Spherical, panoramic lens: Goggles can be worn over prescription glasses
  • Elastic strap in antistatic silicone has an innovative quick adjustment system
  • Indirect ventilation system in the lower part prevents the escape of particulate matter
  • Can be used for prolonged periods as it does not create distortion or fatigue
  • Sterilisable with steam, gamma or beta radiations
Certificeringer: EN 166