DNA transfection kit for protein production, FectoCHO® Expression system
Leverandør: Polyplus-transfection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
101000003 101000011
101000003.EA 7780 DKK
101000003. 101000011.
DNA transfection kit for protein production, FectoCHO® Expression system
Transfektionsreagenser
FectoCHO® expression system consists in the synergistic association of a novel CHO chemically defined medium, FectoCHO® CD expression medium and a powerful transfection reagent, FectoPRO®.
  • Outstanding protein production yields in various CHO cells
  • Fast and effortless CHO cell adaptation
  • Chemically defined kit components
  • Excellent lot to lot reproducibility
  • Cost-effective solution
FectoCHO® CD expression medium allows easy cultivation of various strains of CHO cells such as CHO-K1, CHO-S and ExpiCHO®-S , without the need of an extensive sequential adaptation. FectoPRO®-mediated transient transfection in this chemically defined medium leads to the highest protein and antibody yields ever reached using Transient Gene Expression, up to 4 times higher than with similar systems.
Certificeringer: Polyplus-transfection® has been an ISO 9001 accredited company since 2002. Since then this certification has been successfully renewed every three years.
Bestillingsoplysninger: FectoPRO® transfection reagent and FectoCHO® CD expression medium can be purchased separately.