Magnetic stirring bars, cylindrical, PTFE, fully traceable
Leverandør: COWIE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
FT.001.120 FT.001.130 FT.001.170
COWIFT.001.120EA 9340 DKK
COWIFT.001.120 COWIFT.001.130 COWIFT.001.170
Magnetic stirring bars, cylindrical, PTFE, fully traceable
Omrørere Omrøremagneter
These cylindrical stirring bars feature a smooth, round profile and are ideal for a wide variety of applications.
  • Manufactured from virgin PTFE materials with FDA and USP Class VI approvals
  • With alnico magnet core
  • Chemically inert, non-contaminating and usable to 280 °C
  • Isostatic encapsulation to eliminate cracks and porosity
  • Polished finish to reduce pick-up and cross contamination
Used for stirring and mixing in lquid media and in open and closed systems over a broad temperature range. These bars can be sterilised by chemical or thermal means, but not by gamma radiation.
Certificeringer: ISO 9001:2015.
Emballering: Packed in a grip seal bag.