Powder funnels, SteriWare®
Leverandør: Sampling Systems
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
8111H-200 8111H-200S 8111P-350S 8111P-350 8111H-100 8111H-100S 8111H-50 8111H-50S
SAMP8111H-200EA 999 DKK
SAMP8111H-200 SAMP8111H-200S 300-0533 300-0532 SAMP8111H-100 SAMP8111H-100S SAMP8111H-50 SAMP8111H-50S
Powder funnels, SteriWare®
Tragte Pulvertragte
These single use powder funnels made from chemically resistant, food grade HDPE are ideal for use with powders, creams and liquids.
  • Available as sterile and non sterile versions
  • Manufactured under cleanroom conditions: Can be used straight from the bag
  • Cross-contamination risk is eliminated
  • No cleaning is required before or after sampling
The wide opening at the base of the funnel will help prevent powder blockages.
Certificeringer: Material conforms to FDC and EC standards. Full batch traceability.
Emballering: Individually bagged.