Portable balances, Navigator NV and NVT series
Leverandør: OHAUS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
30456382 30456383 30456384 30456385 30456386 30456387 30456388 30456389 30456390 30456391 30456392 30456393 30456394 30456395 30467835 30467836 30467838 30467837 30615581 30615582 30615583 30615584 30615585 30615586 30615587
611-4459EA 2610 DKK
611-4459 611-4460 611-4464 611-4465 611-4466 611-4467 611-4468 611-4469 611-4470 611-4478 611-4479 611-4480 611-4484 611-4485 611-4666 611-4667 611-4669 611-4668 611-5054 611-5055 611-5056 611-5057 611-5058 611-5059 611-5060
Portable balances, Navigator NV and NVT series
Vægte
Navigator portable balances are perfect for a diverse range of weighing applications in a laboratory or industrial and education settings. Navigator is a powerful balance that is rugged and multifunctional, has fast stabilisation time, overload protection at four times its rated capacity, and is easy to operate.
  • Designed for accuracy and repeatability in essential weighing applications
  • Improved productivity and throughput with fast stabilisation time (less than one second)
  • NV models have been designed to conveniently stack, helping to save storage space