Cryo rack for 48 Cryo.s™ with Datamatrix
Leverandør: Greiner Bio-One
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
803202_20 803270_15 803277_20
479-2148EA 1900 DKK
479-2148 479-2149 GREI803277_20
Cryo rack for 48 Cryo.s™ with Datamatrix
Stativer Stativer til Kryorør
Datamatrix cryo rack for 48 Cryo.s™ storage tubes.
  • Scanning windows in rack bottom for scanning without tube removal
  • Stackable
  • Temperature resistant polypropylene with a temperature range of –196 to +30 °C
Choice of rack configurations:
Rack without lid for 1 ml, 2 ml, 4 ml and 5 ml Cryo.s™ – 30,0 mm high
Rack with small lid for 1 ml and 2 ml Cryo.s™ – 52,5 mm high
Rack with high lid for 4 ml and 5 ml Cryo.s™ – 88,5 mm high
Certificeringer: ANSI/SLAS I-2004 standard footprint rack for ease of automated handling.