Colour collections, RAL DESIGN
Leverandør: Industrial Physics
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VF6621 VF6625
SIMEVF6621EA 19180 DKK
SIMEVF6621 SIMEVF6625
Colour collections, RAL DESIGN
Standarder Farve Referenser og Spektroskopi Standarder
The RAL DESIGN system is used primarily for sophisticated decorative colour design. It is the colour system of choice for architects, designers and advertising agencies.
  • Methodically arranged
  • Colour metric defined colour shades
  • Perfect instrument for individual colour design