Immersion thermostats, DYNEO™ DD
Leverandør: JULABO GmbH
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
9021000 9021000.A 9021000.D
461-1383EA 16020 DKK
461-1383 461-1396 461-1409
Immersion thermostats, DYNEO™ DD
Cirkulatorsystemer Åbne cirkulatorsystemer Styreenheder til termostater
The DYNEO™ DD immersion thermostats ensure high-performance in internal and external applications and have a wide range of modern data communication interfaces. Operation and handling is easy using the large TFT colour display and the state of the art turn and push controller.
  • Easy to switch between internal and external circulation
  • Multi-lingual user interface
  • Powerful and infinitely adjustable pressure pump
  • Integrated programmer for 8×60 program steps
  • Integrated external PT100 connection
  • USB port
Max. flow rate/max. pressure:
22 L/min, 0,6 bar
Certificeringer: Class III (FL) according to DIN 12876-1
Leveringsinformation: Supplied with universal clamp for use in any bath tank up to 50 litres. A range of options, such as units with an RS232 interface or analogue interface are available.