Antibody diluent clear
Leverandør: IMMUNOLOGIC
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BD09-125 BD09-500 BD09-999 S/BD09-20
BD09-125EA 432 DKK
BD09-125 BD09-500 BD09-999 IMMLS/BD09-20
Antibody diluent clear
Elektroforese reagenser Western Blotting-reagenser
Universal IHC Blocking/Diluent is specially formulated to stabilise antibodies and eliminate backgrounds encountered in immunohistochemistry staining procedures. The diluent contains complex proteins and non-protein mixtures which eliminate histostaining background by blocking Fc receptors and prevents non-specific protein-protein interactions.
  • High-quality diluent solution for IHC apllications
Universal IHC Blocking/Diluent can also be used as blocking applied before primary antibodies. If primary antibodies are diluted in this diluent, a blocking step before primary antibodies can generally be ommited. The blocking/diluent solution is supplied in ready-to-use format. It should not be diluted to be effective. The diluent contains no azide, therefore, it can be used to dilute and stabilise HRP-conjugates as well as any other solution that need and stabilising carrier protein, such as post-antibody blocking and poly-AP conjugates.
Certificeringer: Meets CE/IVD standards
Bestillingsoplysninger: Colour version also available (see IMMLS/UD09-20 and similar products)
Leveringsinformation: Shipped with blue ice. Store the products between 2 to 8°C.
Advarsel: Irritating to eyes, respiratory systems and skin