Temperature probes for digital thermometer, DE 305
Leverandør: Amarell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
E908658 E908654 E908656
AMARE908658EA 1480 DKK
AMARE908658 AMARE908654 AMARE908656
Temperature probes for digital thermometer, DE 305
Temperatur prober
These are fast responsing, interchangeable semi conductor probes of NiCr-Ni.
Leveringsinformation: Delivered with 1 m PVC cable.