Qualitative filter papers, grade MN 612 1/4
Leverandør: MACHEREY-NAGEL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
526012 526018 526005 526007 526009 526011 526015 526024 526027 526032 526038 526040 526045 526050
515-9750EA 445 DKK
515-9750 515-9751 MANA526005 MANA526007 MANA526009 MANA526011 MANA526015 MANA526024 MANA526027 MANA526032 MANA526038 MANA526040 MANA526045 MANA526050
Qualitative filter papers, grade MN 612 1/4
Filtre Filtrerpapir Kvalitativt filtrerpapir
Embossed filter papers particularly suited for general laboratory applications.
  • Medium fast filtration
  • Folder filter papers