Mixing measuring cylinders, tall form, class A, borosilicate glass, BLAUBRAND®
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
232408 232428 232420 232438 232448
BRND232408EA 600 DKK
BRND232408 BRND232428 BRND232420 BRND232438 BRND232448
Mixing measuring cylinders, tall form, class A, borosilicate glass, BLAUBRAND®
Cylindere Målecylindre
Borosilicate glass 3.3, class A, conformity certified.
  • Calibrated to contain (TC, In)
  • Hexagonal glass base, PP stopper (except for 612-5028 and 612-5029 octagonal PE stopper)
  • Ring marks at major graduations, marks and inscriptions in high contrast blue enamel
* Height without stopper
Certificeringer: DIN EN ISO 4788
Bestillingsoplysninger: Batch certificate included.