Vial racks for headspace vials
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
22644 22643
REST22644EA 1210 DKK
REST22644 REST22643
Vial racks for headspace vials
Stativer Stativer til Kromatografirør
These vial racks securely hold up to 36 headspace vials.
  • Autoclavable up to 121 °C for 20 minutes
  • Vials lock in rack for convenient one-handed cap removal
  • Easy to clean
  • Stackable even with vials in place
The openings in the bottom of each well facilitate drainage and the alphanumeric indexing on rack simplifies sample identification.