Nitrile gloves, Showa 7555
Leverandør: Showa
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
7555L 7555M 7555S 7555XL 7555XS 7555XXL
BEST7555LEA 307 DKK
BEST7555L BEST7555M BEST7555S BEST7555XL BEST7555XS BEST7555XXL
Nitrile gloves, Showa 7555
Handsker
These powder-free disposable gloves are made from 100% nitrile. They provide protection against the penetration and projection of chemicals.
  • Low-modulus formulation to improve fit and reduce fatigue
  • Chlorinated to offer more comfort, grip and reduce tackiness
  • Textured finish on fingertips to enhance grip
  • Second skin feel, softer texture and easy donning
  • Latex-free to avoid type I and type IV latex allergies
Typical applications include: Aerospace, electronics, food, pharmaceutical and printing.
Certificeringer: EN 455, EN ISO 374-1 Type C (JK), EN ISO 374-5; Cat. III
Food contact approved.