QuEChERS quality control standards for GC-MS analysis
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
33268 33264
REST33268EA 470 DKK
REST33268 REST33264
QuEChERS quality control standards for GC-MS analysis
Standarder Kromatografi Standarder
Pesticide analysis is fast and simple using QuEChERS methods. Use these cost-effective QuEChERS standards for even greaterlab efficiency. Standards are compatible with all major methods, including mini-multiresidue, AOAC, and European procedures.
  • Ready to use for QuEChERS extractions-no dilutions necessary
  • Support for GC and LC with MS, MS/MS, and selective detectors
  • Save time with convenient mixes or make your own blend using our full line of single-component solutions