Capillary columns, OPTIMA® 1301 MS
Leverandør: MACHEREY-NAGEL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
726642.30 726642.60 726640.30 726640.60 726641.30 726641.60 726643.30 726643.60
MANA726642.30EA 10390 DKK
MANA726642.30 MANA726642.60 MANA726640.30 MANA726640.60 MANA726641.30 MANA726641.60 MANA726643.30 MANA726643.60
Capillary columns, OPTIMA® 1301 MS
Kromatografikolonner
Medium polar silarylene GC phase with low bleeding.
  • Ideal for ion trap and quadrupol MS detectors
  • Suitable for sophisticated environmental analysis such as EPA methods for PAH, PCB and pesticides
  • Maximum temperature for isothermal operation 300 °C
  • Maximum temperature for short isotherms in a temperature program 320 °C
The selectivity is analogue to 6% cyanopropyl-phenyl – 94% dimethylpolysiloxane phases.
USP code G43