Molecular sieve A4 (0,4 nm, 4 Å), beads 8 - 12 mesh, Fluka™
Leverandør: Honeywell Chemicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer: Molecular sieve 0.4 nm, Molecular sieves pack 4A, Linde 4A, Sodium aluminium silicate
208604-1KG 208604-5KG
208604-1KGEA 1790 DKK
208604-1KG 208604-5KG
Molecular sieve A4 (0,4 nm, 4 Å), beads 8 - 12 mesh, Fluka™
Molecular sieve A4 (0,4 nm, 4 Å)
Smeltepunkt: 400 °C
Massefylde: 2,0 g/cm³
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00131613
CAS-nummer: 70955-01-0
Bestil nu
Specifikationer Testresultater
Particle Size 8-12 mesh
Grain size 8 - 12 mesh Min. 95 %