Kaliumhydrogensulfat ≥99%, Puriss. p.a., Fluka™
Leverandør: Honeywell Chemicals
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
31260-50KG
31260-1KG
31260-6X1KG
31260-25KG
31260-50KGEA
19910
DKK
31260-50KG
31260-1KG
31260-6X1KG
HONC31260-25KG
Kaliumhydrogensulfat ≥99%, Puriss. p.a., Fluka™
Potassium hydrogen sulphate
Formel:
KHSO₄ Smeltepunkt: 200 °C Massefylde: 2,36 g/cm³ (20 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00011404 CAS-nummer: 7646-93-7 EINECS: 231-594-1 UN nummer: 2509 ADR: 8,II Merck Index: 13,07695 |
Specifikationer Testresultater
Assay | Min. 99 % |
Aluminium (Al) | Max. 0.002 % |
Arsenic (As) | Max. 0.00005 % |
Calcium (Ca) | Max. 0.002 % |
Cadmium (Cd) | Max. 0.0005 % |
Copper (Cu) | Max. 0.0005 % |
Iron (Fe) | Max. 0.0005 % |
Magnesium (Mg) | Max. 0.0005 % |
Sodium (Na) | Max. 0.005 % |
Lead (Pb) | Max. 0.0005 % |
Zinc (Zn) | Max. 0.0005 % |
Chloride (Cl) | Max. 0.0005 % |
Phosphate (PO₄) | Max. 0.0005 % |
Total N | Max. 0.001 % |