Natriumchlorit ~80% (RT), Puriss. p.a., Fluka™
Leverandør: Honeywell Chemicals
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
71388-250G
71388-1KG
71388-250GEA
464
DKK
71388-250G
71388-1KG
Natriumchlorit ~80% (RT), Puriss. p.a., Fluka™
Sodium chlorite
Formel:
ClNaO₂ Smeltepunkt: 180 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00003478 CAS-nummer: 7758-19-2 EINECS: 231-836-6 UN nummer: 1496 ADR: 5.1,II Merck Index: 13,08672 |
Specifikationer Testresultater
Assay | 77.0 - 83.0 % |
Silver (Ag) | Max. 5 ppm |
Aluminium (Al) | Max. 5 ppm |
Barium (Ba) | Max. 5 ppm |
Bismuth (Bi) | Max. 5 ppm |
Calcium (Ca) | Max. 50 ppm |
Cadmium (Cd) | Max. 5 ppm |
Cobalt (Co) | Max. 5 ppm |
Chromium (Cr) | Max. 5 ppm |
Copper (Cu) | Max. 5 ppm |
Iron (Fe) | Max. 5 ppm |
Potassium (K) | Max. 1000 ppm |
Lithium (Li) | Max. 5 ppm |
Magnesium (Mg) | Max. 10 ppm |
Manganese (Mn) | Max. 5 ppm |
Molybdenum (Mo) | Max. 5 ppm |
Nickel (Ni) | Max. 5 ppm |
Lead (Pb) | Max. 5 ppm |
Strontium (Sr) | Max. 5 ppm |
Thallium (Tl) | Max. 5 ppm |
Zinc (Zn) | Max. 5 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...