Acylation derivatisation reagents
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
35621 35623 35625 35620 35622 35624
REST35621EA 2510 DKK
REST35621 REST35623 REST35625 REST35620 REST35622 REST35624
Acylation derivatisation reagents
Kromatografi sorbenter GC Sorbenter
Acylation reagents offer the same advantage available from silylation reagents: creating less polar, more volatile derivatives. In comparison to silylating reagents, the acylating reagents can more readily target highly polar multifunctional compounds, such as carbohydrates and amino acids. In addition, acylating reagents offer the distinct advantage of introducing electron-capturing groups, thus enhancing detectability during analysis.
  • Most commonly used for electron capture detection
  • React with alcohols, amines, and phenols
  • Frequently used for drugs of abuse confirmation