ASTM precision thermometers, stem type
Leverandør: Ludwig Schneider GmbH & CO. KG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1202012S/04/3P 1202015S/04/3P 1202018S/04/3P 1202062S/04/3P 1202091S/04/3P 1202005S/04/3P 1202064S/04/5P 1202120S/04/3P
SCNL1202012S/04/3PEA 4340 DKK
SCNL1202012S/04/3P SCNL1202015S/04/3P SCNL1202018S/04/3P SCNL1202062S/04/3P SCNL1202091S/04/3P SCNL1202005S/04/3P SCNL1202064S/04/5P SCNL1202120S/04/3P
ASTM precision thermometers, stem type
Termometre Analoge termometre
These precision thermometers are available with DAkks calibration certificate.
  • Round, white back capillary form
Blue filling, stem type
Emballering: Packed individualy in a square plastic sleeve.