Single-component explosives solutions
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
31452 31453 31498 31499 31600 31601 31602 31612 31657 31658 31659 31660 31661 31662 31663 31664 31665 31666 31667 31668 31669 31670 31671 31821 33901
REST31452EA 388 DKK
REST31452 REST31453 REST31498 REST31499 REST31600 REST31601 REST31602 REST31612 REST31657 REST31658 REST31659 REST31660 REST31661 REST31662 REST31663 REST31664 REST31665 REST31666 REST31667 REST31668 REST31669 REST31670 REST31671 REST31821 REST33901
Single-component explosives solutions
Standarder Økologiske standarder
Single-component explosives solutions supports US department of defense base closures and remediation.
  • Mixtures and singles supporting LC US EPA method 8330
  • Mixtures and singles supporting GC-ECD US EPA method 8095
  • Internal standards and surrogates to support both methods
These materials support nitroaromatic, nitramine, and nitroester analyses by GC-ECD (Method 8095). Compounds listed are explosives, manufacturing intermediates, or degradation products. Method 8095 mixtures contains the components at concentration ratios appropriate for ECD.