Cleaner and degreaser, SURE®
Leverandør: Diversey
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
100894246 100894247
141-1586EA 477 DKK
141-1586 141-1587
Cleaner and degreaser, SURE®
Renseopløsninger Detergenter og Overflade Rengøringsmidler
Concentrated heavy-duty degreaser for cleaning of all heavily soiled surfaces in kitchens like counter tops, stove top and kitchen appliance surfaces. It is also suitable for heavy-duty cleaning of floors.
  • No fragrance or dye
  • Contains plant-based ingredients from renewable sources
  • Decomposed by natural processes
  • No environmental hazard classifications, no safety warnings for product in use
  • Concentrated formulation gives excellent economy in use
  • Powerful formulation cuts through heavy grease leaving clean and shiny surfaces
  • Reduced environmental impact
SURE® cleaner and degreaser is part of a unique range of plant-based, cleaning products that deliver the professional cleaning results you need - while being safe for people and kind to the environment. SURE® natural cleaning products provide safety and sustainability without compromising on performance.
Bottles/cans suitable for recycling.
Certificeringer: OECD 301B.
EU-Ecolabel (EU: NL/020/045)
Cradle to cradle certification at gold level.