Chiral HPLC columns, NUCLEOCEL DELTA
Leverandør: MACHEREY-NAGEL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
720445.46 720446.46 720450.46 720451.46 721185.30 721186.30
MANA720445.46EA 37890 DKK
MANA720445.46 MANA720446.46 MANA720450.46 MANA720451.46 MANA721185.30 MANA721186.30
Chiral HPLC columns, NUCLEOCEL DELTA
Kromatografikolonner
Cellulose-modified silica phases for HPLC enantiomer separation.
  • Base material: silica gel
  • Chiral selector: cellulose tris-(3,5-dimethylphenylcarbamate)
  • Phases for normal phase or reversed phase mode
NUCLEOCEL DELTA is recommended for enantiomer separation methods in reference to a successful application note and for chiral method developments of pharmaceutically active compounds, pollutants, food ingredients, catalysts, and bioorganic compounds.
EC guard columns are used for analytical EC columns with the Column Protection System guard column holder (554-5851).