Body showers
Leverandør: BROEN-LAB
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
17581009 17582009 17580009 17587409
BROE17581009EA 6170 DKK
BROE17581009 BROE17582009 118-0551 BROE17587409
Body showers
Førstehjælpsudstyr Brusere
Body shower for exposed pipework with self-draining head. In brass with chemical resistant BROEN-LAB Polycoat.
  • Professional solutions for employee safety
  • Easy to mount
  • Anti-limescale design and material
  • Improved functionality with long lifetime
  • Strong wall flange with 6 holes for wall mounted models with exposed piping
Min. working pressure: 1 bar
Certificeringer: EN 15154-1
Bestillingsoplysninger: Supporting wall brackets for mounting must be ordered separately.