Storage drums
Leverandør: Sampling Systems
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
A126-05 A126-10 A126-15 A126-01 A126-02 A126-03
SAMPA126-05EA 6210 DKK
SAMPA126-05 SAMPA126-10 SAMPA126-15 300-0620 300-0621 300-0622
Storage drums
Tønder, fustager, trommer
These drums are used for the storage and processing of powders and liquids.
  • Open top pharmaceutical grade drums with lids
  • Made of 316L stainless steel body and lid
  • Supplied with side handles
The robust, hygienic design means they are suitable for the most demanding applications. These drums are finished to a bright polish finish with all the welds ground and polished.
Leveringsinformation: Supplied with lid, clamp and O-ring.