Aluminiumoxid aktiveret, neutral Brockmann Grade I ~150 mesh, Fluka™

Leverandør: Honeywell Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

199974-1KG 199974-5KG 199974-25KG
199974-1KGEA 1240 DKK
199974-1KG 199974-5KG HONC199974-25KG
Aluminiumoxid aktiveret, neutral Brockmann Grade I ~150 mesh, Fluka™
Aluminium oxide activated neutral
Formel: Al₂O₃
Kogepunkt: 2,980 °C (1.013 hPa)
Massefylde: 3,960 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003424
CAS-nummer: 1344-28-1
EINECS: 215-691-6

Bestil nu

Specifikationer Testresultater

Brockmann Number (Activity) I (val.ref) complying
pH (10 % susp., 20°C) 6.5 - 7.5
Spez. surface (BET) 135 - 165 m²/g
Fe (as Fe₂O₃) Max. 0.03 %
Na (as Na₂O) Max. 0.4 %
Silicon (as SiO₂) Max. 0.03 %
Chloride (Cl) Max. 0.2 %
Grain size 0.04-0.30 mm Min. 95 %
Pore Size 58 +� pore size


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR