Polypropylene Laboratory Labels, M211 and M210
Leverandør: Brady
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
121014 121015 121016
116-0628EA 272 DKK
116-0628 116-0629 116-0630
Polypropylene Laboratory Labels, M211 and M210
Etiketter
Polypropylene labelling tapes for vials, tubes, slides and well plates.
  • Matt finish
  • For freezer or liquid nitrogen applications
B-7425 must be applied prior to being frozen.