Self-Laminating Vinyl Wire and Cable Labels, M211 and M210
Leverandør: Brady
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
110927 110928
116-0623EA 336 DKK
116-0623 116-0624
Self-Laminating Vinyl Wire and Cable Labels, M211 and M210
Etiketter
Self-laminating vinyl labelling tape with clear wraparound size of 9,52 mm.
  • Excellent abrasion, smudge, water and oil resistance
  • Self-extinguishing
  • Matt finish
  • For smooth and textured/rough surfaces