General purpose thermometers, simple type
Leverandør: Amarell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
G11500-FL G11502-FL G11504-FL G11506-FL G11580-FL
AMARG11500-FLEA 65.3 DKK
AMARG11500-FL AMARG11502-FL AMARG11504-FL AMARG11506-FL AMARG11580-FL
General purpose thermometers, simple type
Termometre Analoge termometre
These simple type general purpose thermometers have a solid stem with ring at top.
  • Durable pigment, low cost type
  • Total immersion, Ø 6,0 to 7,0 mm
Emballering: Packed in a transparent square plastic case.