AQUAMETRIC titrants for volumetric Karl Fischer titrations
Leverandør: PanReac AppliChem
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
285815.1612
285815.1611
285816.1611
APPC285815.1612EA
3600
DKK
APPC285815.1612
APPC285815.1611
APPC285816.1611
AQUAMETRIC titrants for volumetric Karl Fischer titrations
Karl Fischer titranter og reagenser
- Use with AQUAMETRIC solvent
1 ml of Titrant 5 corresponds to: min. 5,0 mg of H₂O (20°C) at time of manufacture.
1 ml of Titrant 2 corresponds to: min. 2,0 mg of H₂O (20°C) at time of manufacture.