Thermal transfer printer labels, type B-483, ToughBond™
Leverandør: Brady
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
031331 031336 031335 031337 031330
BRDY031331EA 1800 DKK
BRDY031331 BRDY031336 BRDY031335 BRDY031337 BRDY031330
Thermal transfer printer labels, type B-483, ToughBond™
Etiketter
Printable, ultra aggressive polyester labels for thermal transfer printers.
  • Die-cut labels are supplied on a roll
  • Contains some adhesive backed wire marking materials, and also labels that are pre-printed with headers or templates
  • Supplied with the ribbon seperate, not in the same enclosure as the tape/sleeves
This material is UL and CSA approved with its respective ribbon. See the recommended ribbon series for the appropriate ribbon to use.